Anti-HDAC10 Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Supplier: Bioss

BS-2893R-A488
10355-002EA 811.1 USD
10355-002
Anti-HDAC10 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antibodies
Responsible for the deacetylation of lysine residues on the N-terminal part of the core histones (H2A, H2B, H3 and H4). Histone deacetylation gives a tag for epigenetic repression and plays an important role in transcriptional regulation, cell cycle progression and developmental events. Histone deacetylases act via the formation of large multiprotein complexes.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: HDAC10
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR