Anti-V5 Tag Rabbit Polyclonal Antibody

Supplier: Bioss
BS-2109R
10337-314EA 573.73 USD
10337-314
Anti-V5 Tag Rabbit Polyclonal Antibody
Antibodies
The V5 epitope tag is derived from a small epitope (Pk) present on the P and V proteins of the paramyxovirus of simian virus 5 (SV5). The V5 tag is usually used with all 14 amino acids, although it has also been used with a shorter 9 amino acid sequence.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: V5 Tag
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Tags
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR