Anti-Neuronal thread protein AD7c-NTP Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

BS-2105R-FITC
10337-262EA 592.03 USD
10337-262
Anti-Neuronal thread protein AD7c-NTP Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
This gene is unusual in that its coding sequence is derived almost entirely from a cluster of different Alu repeat sequences. However, the mRNA and the encoded protein have been shown to be expressed in neurons, and overexpressed in brains with Alzheimer's disease. In vitro studies also demonstrated that abnormal expression of this gene promoted neuritic sprouting and cell death, associated with dementia in Alzheimer's disease.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: Neuronal thread protein AD7c-NTP
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR