Anti-Neuronal thread protein AD7c-NTP Rabbit Polyclonal Antibody (Biotin)

BS-2105R-BIO
10337-252EA 529.29 USD
10337-252
Anti-Neuronal thread protein AD7c-NTP Rabbit Polyclonal Antibody (Biotin)
This gene is unusual in that its coding sequence is derived almost entirely from a cluster of different Alu repeat sequences. However, the mRNA and the encoded protein have been shown to be expressed in neurons, and overexpressed in brains with Alzheimer's disease. In vitro studies also demonstrated that abnormal expression of this gene promoted neuritic sprouting and cell death, associated with dementia in Alzheimer's disease.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500

Type: Primary
Antigen: Neuronal thread protein AD7c-NTP
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR