Anti-PRKDC Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Supplier: Bioss
BS-2092R-HRP
10337-010EA 757.96 USD
10337-010
Anti-PRKDC Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antibodies
DNA PK is the key component of the non-homologous end-joining (NHEJ) pathway of DSB repair in mammalian cells. DNA PK consists of a heterodimeric DNA-binding subunit (Ku70/80) and an approximately 465 kDa catalytic subunit (DNA PKcs). DNA PKcs is a serine/threonine protein kinase whose activity is greatly stimulated by its recruitment to DNA breaks by the Ku heterodimer.

Recommended Dilutions: IHC-P: 1:100-500

Type: Primary
Antigen: PRKDC
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR