Anti-PRKDC Rabbit Polyclonal Antibody (Cy5.5®)

Supplier: Bioss
BS-2092R-CY5.5
10337-004EA 746.42 USD
10337-004
Anti-PRKDC Rabbit Polyclonal Antibody (Cy5.5®)
Antibodies
DNA PK is the key component of the non-homologous end-joining (NHEJ) pathway of DSB repair in mammalian cells. DNA PK consists of a heterodimeric DNA-binding subunit (Ku70/80) and an approximately 465 kDa catalytic subunit (DNA PKcs). DNA PKcs is a serine/threonine protein kinase whose activity is greatly stimulated by its recruitment to DNA breaks by the Ku heterodimer.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: PRKDC
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR