Anti-TNFRSF10C Rabbit Polyclonal Antibody (Cy7®)

Supplier: Bioss

BS-2083R-CY7
10336-254EA 811.1 USD
10336-254
Anti-TNFRSF10C Rabbit Polyclonal Antibody (Cy7®)
Antibodies
Receptor for the cytotoxic ligand TRAIL. Lacks a cytoplasmic death domain and hence is not capable of inducing apoptosis. May protect cells against TRAIL mediated apoptosis by competing with TRAIL-R1 and R2 for binding to the ligand.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: TNFRSF10C
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR