Anti-TNF Rabbit Polyclonal Antibody (Cy3®)

Supplier: Bioss

BS-2081R-CY3
10336-288EA 811.1 USD
10336-288
Anti-TNF Rabbit Polyclonal Antibody (Cy3®)
Antibodies
Cytokine that binds to TNFRSF1A/TNFR1 and TNFRSF1B/TNFBR. It is mainly secreted by macrophages and can induce cell death of certain tumor cell lines. It is potent pyrogen causing fever by direct action or by stimulation of interleukin-1 secretion and is implicated in the induction of cachexia, Under certain conditions it can stimulate cell proliferation and induce cell differentiation. The TNF intracellular domain (ICD) form induces IL12 production in dendritic cells.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: TNF
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR