Anti-alpha-L-arabinofuranosidase Rabbit Polyclonal Antibody (Cy3®)

Supplier: Bioss

BS-2045R-CY3
10336-000EA 811.1 USD
10336-000
Anti-alpha-L-arabinofuranosidase Rabbit Polyclonal Antibody (Cy3®)
Antibodies
no data available.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: [delete]
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Plant
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR