Anti-alpha-L-arabinofuranosidase Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Supplier: Bioss

BS-2045R-A350
10335-468EA 811.1 USD
10335-468
Anti-alpha-L-arabinofuranosidase Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antibodies
no data available.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: [delete]
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Plant
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR