Anti-CK18 Rabbit Polyclonal Antibody (PE (Phycoerythrin))

Supplier: Bioss

BS-2043R-PE
10335-996EA 828.8 USD
10335-996
Anti-CK18 Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Antibodies
Involved in the uptake of thrombin-antithrombin complexes by hepatic cells (By similarity). When phosphorylated, plays a role in filament reorganization. Involved in the delivery of mutated CFTR to the plasma membrane. Together with KRT8, is involved in interleukin-6 (IL-6)-mediated barrier protection.

Recommended Dilutions: Flow Cytometry: 1:20-100

Type: Primary
Antigen: CK18
Clonality: Polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR