Anti-PRCP Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Supplier: Bioss

BS-1873R-A488
10331-438EA 811.1 USD
10331-438
Anti-PRCP Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antibodies
Cleaves C-terminal amino acids linked to proline in peptides such as angiotensin II, III and des-Arg9-bradykinin. This cleavage occurs at acidic pH, but enzymatic activity is retained with some substrates at neutral pH.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: PRCP
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR