Anti-GPR78 Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Supplier: Bioss

BS-1807R-A350
10329-454EA 811.1 USD
10329-454
Anti-GPR78 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antibodies
Orphan receptor. Displays a significant level of constitutive activity. Its effect is mediated by G(s)-alpha protein that stimulate adenylate cyclase, resulting in an elevation of intracellular cAMP.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: GPR78
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR