Anti-IL6RA Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Supplier: Bioss

BS-1805R-A488
10329-408EA 811.1 USD
10329-408
Anti-IL6RA Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antibodies
Part of the receptor for interleukin 6. Binds to IL6 with low affinity, but does not transduce a signal. Signal activation necessitate an association with IL6ST. Activation may lead to the regulation of the immune response, acute-phase reactions and hematopoiesis.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: IL6RA
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR