Anti-NPTN Rabbit Polyclonal Antibody (Cy7®)

Supplier: Bioss

BS-1801R-CY7
10329-186EA 811.1 USD
10329-186
Anti-NPTN Rabbit Polyclonal Antibody (Cy7®)
Antibodies
NPTN is a type I transmembrane protein belonging to the Ig superfamily. The protein is believed to be involved in cell-cell interactions or cell-substrate interactions. The alpha and beta transcripts show differential localization within the brain.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: NPTN
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR