Anti-GRIA3 Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Supplier: Bioss

BS-1799R-A488
10329-100EA 811.1 USD
10329-100
Anti-GRIA3 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antibodies
Receptor for glutamate that functions as ligand-gated ion channel in the central nervous system and plays an important role in excitatory synaptic transmission. L-glutamate acts as an excitatory neurotransmitter at many synapses in the central nervous system. Binding of the excitatory neurotransmitter L-glutamate induces a conformation change, leading to the opening of the cation channel, and thereby converts the chemical signal to an electrical impulse. The receptor then desensitizes rapidly and enters a transient inactive state, characterized by the presence of bound agonist. In the presence of CACNG4 or CACNG7 or CACNG8, shows resensitization which is characterized by a delayed accumulation of current flux upon continued application of glutamate.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: GLUR3/GRIA 3
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR