Anti-CXCL5 Rabbit Polyclonal Antibody (Cy5.5®)

BS-1793R-CY5.5
10328-384EA 609.8 USD
10328-384
Anti-CXCL5 Rabbit Polyclonal Antibody (Cy5.5®)
May participate in the recruitment of inflammatory cells by injured or infected tissue. GCP-2(1-78) and, more potent, GCP-2(9-78) attract neutrophils and are involved in neutrophil activation.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CXCL5
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR