Anti-STAT6 Rabbit Polyclonal Antibody (Cy5®)

Supplier: Bioss

BS-1791R-CY5
10328-966EA 811.12 USD
10328-966
Anti-STAT6 Rabbit Polyclonal Antibody (Cy5®)
Antibodies
Carries out a dual function: signal transduction and activation of transcription. Involved in IL4/interleukin-4- and IL3/interleukin-3-mediated signaling.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: STAT6
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR