Anti-IL12A Rabbit Polyclonal Antibody (Cy5®)

Supplier: Bioss

BS-1789R-CY5
10328-838EA 811.1 USD
10328-838
Anti-IL12A Rabbit Polyclonal Antibody (Cy5®)
Antibodies
Cytokine that can act as a growth factor for activated T and NK cells, enhance the lytic activity of NK/lymphokine-activated Killer cells, and stimulate the production of IFN-gamma by resting PBMC.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: IL12A
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR