Anti-ECE2 Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Supplier: Bioss

BS-1753R-A488
10327-448EA 811.1 USD
10327-448
Anti-ECE2 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antibodies
Converts big endothelin-1 to endothelin-1. Also involved in the processing of various neuroendocrine peptides, including neurotensin, angiotensin I, substance P, proenkephalin-derived peptides, and prodynorphin-derived peptides. May limit beta-amyloid peptide accumulation in brain. May also have methyltransferase activity.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ECE2
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR