Anti-CFH Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Supplier: Bioss
BS-1745R-A488
10327-456EA 811.1 USD
10327-456
Anti-CFH Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antibodies
Factor H functions as a cofactor in the inactivation of C3b by factor I and also increases the rate of dissociation of the C3bBb complex (C3 convertase) and the (C3b)NBB complex (C5 convertase) in the alternative complement pathway.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CFH
Clonality: Polyclonal
Clone: 0
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR