Anti-ZBED3 Rabbit Polyclonal Antibody (Cy5®)

Supplier: Bioss

BS-13553R-CY5
10300-628EA 811.1 USD
10300-628
Anti-ZBED3 Rabbit Polyclonal Antibody (Cy5®)
Antibodies
ZBED3

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ZBED3
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Rabbit
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR