Anti-ZBED2 Rabbit Polyclonal Antibody (Cy7®)

Supplier: Bioss

BS-13552R-CY7
10300-366EA 811.1 USD
10300-366
Anti-ZBED2 Rabbit Polyclonal Antibody (Cy7®)
Antibodies
ZBED2

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ZBED2
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR