Anti-ZBED2 Rabbit Polyclonal Antibody (Cy5.5®)

13552R-CY5.5
10227-294EA 559.62 USD
10227-294
Anti-ZBED2 Rabbit Polyclonal Antibody (Cy5.5®)
ZBED2

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ZBED2
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR