Anti-ZAR1L Rabbit Polyclonal Antibody (Cy3®)

Supplier: Bioss

BS-13550R-CY3
10302-798EA 811.1 USD
10302-798
Anti-ZAR1L Rabbit Polyclonal Antibody (Cy3®)
Antibodies
This gene encodes a member of the ZAR1 family that is predominantly expressed in oocytes and early embryos. The protein may function as an RNA regulator in early embryos.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ZAR1L
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR