Anti-ZAR1 Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Supplier: Bioss

BS-13549R-A350
10302-766EA 811.1 USD
10302-766
Anti-ZAR1 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antibodies
he female gamete, the oocyte, serves the distinct purpose of transmitting the maternal genome and other maternal factors critical for postovulation events. Oocytes have diverse functions in ovarian folliculogenesis, fertilization, and embryogenesis. ZAR1 is an oocyte-specific gene that appears to function at the oocyte-to-gamete transition

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ZAR1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR