Anti-ZAN Rabbit Polyclonal Antibody (Cy5®)

Supplier: Bioss

BS-13547R-CY5
10302-756EA 811.1 USD
10302-756
Anti-ZAN Rabbit Polyclonal Antibody (Cy5®)
Antibodies
This gene encodes a sperm membrane protein that binds the zona pellucida of the egg in a species-specific manner. The encoded protein may be involved in signaling or gamete recognition. Alternate transcriptional splice variants, encoding different isoforms, have been characterized. [provided by RefSeq, Jul 2008].

Recommended Dilutions: Flow Cytometry: 1:20-100; IF(IHC-P): 1:50-200

Type: Primary
Antigen: ZAN
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR