Anti-ZAK/HCCS4 Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Supplier: Bioss

BS-13546R-A555
10300-596EA 811.1 USD
10300-596
Anti-ZAK/HCCS4 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antibodies
Stress-activated component of a protein kinase signal transduction cascade. Regulates the JNK and p38 pathways. Pro-apoptotic. Role in regulation of S and G2 cell cycle checkpoint by direct phosphorylation of CHEK2. Isoform 1, but not isoform 2, causes cell shrinkage and disruption of actin stress fibers. Isoform 1 may have role in neoplastic cell transformation and cancer development. Isoform 1, but not isoform 2, phosphorylates histone H3 at 'Ser-28'.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ZAK/HCCS4
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR