|
![]() |
ORDER
SPECIFICATIONS
Antigen Symbol | GUCY1A2 |
Antigen Name | Guanylyl Cyclase alpha 2 |
Reactivity | Human, Rat, Mouse |
Conjugation | HRP (Horseradish Peroxidase) |
Clonality | Polyclonal |
Antibody Type | Primary |
Host | Rabbit |
Isotype | IgG |
Western Blot | Yes |
ImmunoChemistry | Yes |
Antigen Synonyms | GUC1A2, alpha-2 chain, GCS-alpha-2, GUCY1A2, GUCSA2, Guanylate cyclase soluble |
Gene ID | 2977 |
Concentration | 1 μg/μl |
Cross Adsorption | No |
Purification | Purified by Protein A |
Form | liquid |
Storage Buffer | Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% Gentamicin. Store at 4°C for 12 months. |
Storage Temperature | Store at 4°C for 12 months |
Shipping Temperature | 4°C |