Anti-BECN1 Rabbit Polyclonal Antibody (Cy3®)

Supplier: Bioss

BS-1353R-CY3
10300-292EA 811.1 USD
10300-292
Anti-BECN1 Rabbit Polyclonal Antibody (Cy3®)
Antibodies
Plays a central role in autophagy. Required for the abcission step in cytokinesis. May play a role in antiviral host defense. Protects against infection by a neurovirulent strain of Sindbis virus.

Recommended Dilutions: IF(IHC-P): 1:50-200; IF(ICC)(1:50-200

Type: Primary
Antigen: BECN1
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR