Anti-GPR34 Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Supplier: Bioss

BS-13532R-A647
10300-162EA 811.1 USD
10300-162
Anti-GPR34 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antibodies
G protein-coupled receptors (GPRs), also known as seven transmembrane receptors, heptahelical receptors or 7TM receptors, comprise a superfamily of proteins that play a role in many different stimulus-response pathways. GPR signaling is an evolutionarily ancient mechanism used by all eukaryotes to sense environmental stimuli and mediate cell-cell communication. G protein-coupled receptors translate extracellular signals into intracellular signals (G protein activation) and they respond to a variety of signaling molecules, such as hormones and neurotransmitters. GPR34 is a 381 amino acid protein belonging to the G-protein coupled receptor 1 family. Widely expressed, GPR34 is localized to the cell membrane.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: GPR34
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR