Anti-MCL1 Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Supplier: Bioss
BS-1352R-A647
10300-142EA 757.96 USD
10300-142
Anti-MCL1 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antibodies
Involved in the regulation of apoptosis versus cell survival, and in the maintenance of viability but not of proliferation. Mediates its effects by interactions with a number of other regulators of apoptosis. Isoform 1 inhibits apoptosis. Isoform 2 promotes apoptosis.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: MCL1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR