Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-GPR3 Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Supplier: Bioss
G protein-coupled receptor 3 (GPR3), also designated ACCA orphan receptor, is a 330 amino acid member of the G protein-coupled receptor 1 family. The function of GPR3 is mediated by G proteins which activate adenylate cyclase. GPR3 is a mutli-pass membrane protein that is located on the cellular membrane of cells and is detected at low levels in the eye, kidney, lung, ovary and testis. GPR3 is most highly expressed in the central nervous system, where it stimulates the production of cAMP, leading to neurite outgrowth and myelin inhibition. In oocytes, this control over cAMP production can halt meiosis and prevent progesterone-induced meiotic maturation. Mice deficient for GPR3 are able to reproduce but have no control over the oocyte maturation process, which results in nondeveloping early embryos and fragmented oocytes as the mice age.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: GPR3
Clonality: Polyclonal
Conjugation: Alexa Fluor® 488
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Log in to see your contract pricing and availability.
  Remember me on this device


Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
100 µL BS-13529R-A488 10300-110 Each (100µl) Retrieving Restricted

Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Requires a maximum 2 day delivery.

restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page


Antigen Symbol GPR3
Antigen Name G Protein-coupled Receptor 3
Reactivity Mouse, Human, Rat
Conjugation Alexa Fluor® 488
Clonality Polyclonal
Antibody Type Primary
Host Rabbit
Isotype IgG
ImmunoChemistry Yes
Antigen Synonyms ACCA orphan receptor, ACCA, G-protein coupled receptor 3, GPR3
Gene ID 2827
Concentration 1 μg/μl
Cross Adsorption No
Purification Purified by Protein A
Form liquid
Storage Buffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Storage Temperature Store at 4°C for 12 months
Shipping Temperature 4°C
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty