Anti-GPR146 Rabbit Polyclonal Antibody (Cy5®)

Supplier: Bioss

BS-13519R-CY5
10299-922EA 811.1 USD
10299-922
Anti-GPR146 Rabbit Polyclonal Antibody (Cy5®)
Antibodies
G protein-coupled receptors (GPRs), also known as seven transmembrane receptors, heptahelical receptors or 7TM receptors, comprise a superfamily of proteins that play a role in many different stimulus-response pathways. G protein coupled receptors translate extracellular signals into intracellular signals (G protein activation) and they respond to a variety of signaling molecules, such as hormones and neurotransmitters. GPR146 (G protein-coupled receptor 146), also known as PGR8, is a 333 amino acid multi-pass transmembrane protein that belongs to the G-protein coupled receptor 1 family. Characterized as an orphan receptor for which its endogenous ligand has yet to be identified, GPR146 is thought to play a role in signaling events throughout the cell.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: GPR146
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR