Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-GPR132 Rabbit Polyclonal Antibody (Cy5®)

Supplier: Bioss
BS-13508R-CY5
G2A (for G2 accumulation) is a seven transmembrane G protein-coupled receptor that is upregulated in response to DNA damage and stress (1). G2A is predominantly expressed in hematopoietic tissues and in hematopoietic stem cells, and it is more highly detected in pro-B cells, while lower expression is observed in immature B cells and pre-B cells (1,2). G2A is expressed throughout T cell maturation, and it is further increased in response to T-cell activation (3). Ectopic expression of a G2A fusion protein in NIH/3T3 fibroblasts induces a cell cycle arrest that is consistent with a block at the G2/M transition (1,4). G2A is also able to attenuate the proliferative effects of Bcr-Abl, a chimeric tyrosine kinase oncogene, suggesting that G2A possesses anti-oncogenic properties (5). The amino acid sequence of G2A contains a destruction box motif that is consistently observed in cyclins, where it is required for ubiquitination and proteolytic degradation (6).

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: GPCR G2A
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human




Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
100 µL BS-13508R-CY5 10299-514 Each (100µl) Retrieving Restricted

Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Requires a maximum 2 day delivery.

restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen Symbol GPR132
Antigen Name G Protein-coupled Receptor 132
Reactivity Human
Conjugation Cy5®
Clonality Polyclonal
Antibody Type Primary
Host Rabbit
Isotype IgG
ImmunoChemistry Yes
Antigen Synonyms G2A
Gene ID 29933
Concentration 1 μg/μl
Cross Adsorption No
Purification Purified by Protein A
Form liquid
Storage Buffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Storage Temperature Store at 4°C for 12 months
Shipping Temperature 4°C
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Select Product:

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty