Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-C5AR2 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Supplier: Bioss
Anaphylatoxin, a chemotactic and inflammatory peptide, is a receptor for C5a anaphylatoxin chemotactic receptor (C5L2), also designated G protein-coupled receptor 77. C5L2 is an integral membrane protein that belongs to the Gprotein-coupled receptor 1 family. C5L2 weakly couples to Gi-mediated signaling pathways and is widely expressed in the hippocampus, hypothalamus, pons, frontal cortex and liver. It is coexpressed with the C5a receptor (C5AR) on polymorphonuclear neutrophils and may modulate the activity of C5AR. The function of the C5a receptors, particularly that of C5L2, is still unclear. Research suggests that C5L2 may be required for the in vitro release of high mobility group box 1 protein (HMGB1).

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500

Type: Primary
Antigen: C5AR2
Clonality: Polyclonal
Conjugation: HRP (Horseradish Peroxidase)
Host: Rabbit
Isotype: IgG1
Reactivity: Human

Log in to see your contract pricing and availability.
  Remember me on this device


Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
100 µL BS-13506R-HRP 10299-646 Each (100µl) Retrieving Restricted

Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Requires a maximum 2 day delivery.

restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page


Antigen Symbol C5AR2
Antigen Name Complement Component 5a receptor 2
Reactivity Human
Conjugation HRP (Horseradish Peroxidase)
Clonality Polyclonal
Antibody Type Primary
Host Rabbit
Isotype IgG
Western Blot Yes
ImmunoChemistry Yes
Antigen Synonyms GPF77, GPR77, C5L2
Gene ID 27202
Concentration 1 μg/μl
Cross Adsorption No
Purification Purified by Protein A
Form liquid
Storage Buffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% Gentamicin. Store at 4°C for 12 months.
Storage Temperature Store at 4°C for 12 months
Shipping Temperature 4°C
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty