Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-GPR151 Rabbit Polyclonal Antibody (PE (Phycoerythrin))

Supplier: Bioss
GPCR2037 is a G protein-coupled receptor that undergoes weak activation by Galanin and is most abundant in the central nervous system (CNS), where it appears to be critical for development. During embryonal development the expression of GPCR2037 is widespread in the nervous system (dorsal thalamus, striatum, locus coeruleus and hindbrain nuclei). GPCR2037 in the CNS of 7- and 15-day-old mouse embryos can localize to the habenular complex. Low levels of GPCR2037 are detectable in testis, liver, kidney and stomach. In addition to GPCR2037, Galanin mediates its effects through receptor subtypes GALR1, 2 and 3. Galanin ligand exerts anxiolytic actions via GALR receptors under conditions of high stress. Galanin coexists with norepinephrine and serotonin in neural systems that mediate emotion.

Recommended Dilutions: Flow Cytometry: 1:20-100

Type: Primary
Antigen: GPR 151
Clonality: Polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat



Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
100 µL BS-13505R-PE 10299-624 Each (100µl) Retrieving Restricted
restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen Symbol GPR151
Antigen Name G Protein-coupled Receptor 151
Reactivity Mouse, Rat, Human
Conjugation PE (Phycoerythrin)
Clonality Polyclonal
Antibody Type Primary
Host Rabbit
Isotype IgG
Flow Cytometry Yes
Antigen Synonyms PGR7, GALRL, GALR4, GPCR
Gene ID 134391
Concentration 1 μg/μl
Cross Adsorption No
Purification Purified by Protein A
Form liquid
Storage Buffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Storage Temperature Store at 4°C for 12 months
Shipping Temperature 4°C
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Select Product:

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty