Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Global Web is currently experiencing issues, technical teams are working to correct the problem.

Anti-GPBP1L1 Rabbit Polyclonal Antibody (Cy3®)

Supplier: Bioss
Transcription factors are required for the initiation of transcription. They regulate transcription by binding to DNA at specific nucleotide sequences within promoters and enhancers. Transcription factors, which may also bind to RNA polymerase or to other transcription factors, are involved in the preinitiation complex formation. Upstream transcription factors and inducible transcription factors bind upstream of the initiation site to repress or stimulate transcription. Upstream factors are unregulated, while inducible factors require inhibition or activation. GPBP1L1 (GC-rich promoter binding protein 1-like 1), also known as vasculin-like protein 1 or SP192, is a 474 amino acid protein belonging to the vasculin family. Localizing to nucleus, GPBP1L1 may function as a transcription factor. The gene encoding GPBP1L1 maps to human chromosome 1p34.1 and mouse chromosome 4 D1.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: GPBP1L1
Clonality: Polyclonal
Conjugation: Cy3®
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Log in to see your contract pricing and availability.
  Remember me on this device


Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
100 µL BS-13504R-CY3 10299-442 Each (100µl) Retrieving Restricted

Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Requires a maximum 2 day delivery.

restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page


Antigen Symbol GPBP1L1
Antigen Name Gc-Rich Promoter Binding Protein 1-Like 1
Reactivity Mouse, Rat, Human
Conjugation Cy3®
Clonality Polyclonal
Antibody Type Primary
Host Rabbit
Isotype IgG
ImmunoChemistry Yes
Antigen Synonyms Vasculin-like protein 1., GPBP1L1, SP192, GPBL1_HUMAN, GC-rich promoter-binding protein 1-like 1
Gene ID 60313
Concentration 1 μg/μl
Cross Adsorption No
Purification Purified by Protein A
Form liquid
Storage Buffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Storage Temperature Store at 4°C for 12 months
Shipping Temperature 4°C
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty