Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-GPBP1 Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Supplier: Bioss
BS-13503R-A488
Vascular wall-linked protein, or Vasculin, is primarily expressed in the arterial wall and in plasma. It is also differentially expressed in human atherogenesis. Alternative splicing of exon 3 of the Vasculin gene produces three variants. Vasculin binds to and activates the minimal self-sufficient promoter element (MSPE) of the mouse Ada gene promoter and binds to and partially suppresses the GC-rich promoter of the nonhomologous human TOP2A gene promoter. It acts as a nuclear factor that can form complexes with TATA-binding proteins, transcription factors TFIIB and TFIIF, RNA polymerase II and p300. The regulated expression of Vasculin in plaques suggests that it may be involved in atherogenesis, and its presence in plasma may implicate Vasculin as a marker for atherosclerosis.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: GPBP1/Vasculin
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat




 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
100 µL BS-13503R-A488 10299-604 Each (100µl) Retrieving Restricted

Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Shipping duration cannot exceed 2 days.

restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen Symbol GPBP1
Antigen Name GC-rich promoter binding protein 1
Reactivity Human, Rat, Mouse
Conjugation Alexa Fluor® 488
Clonality Polyclonal
Antibody Type Primary
Host Rabbit
Isotype IgG
ImmunoChemistry Yes
Antigen Synonyms SSH6, GPBP, VASCULIN
Gene ID 65056
Concentration 1 μg/μl
Cross Adsorption No
Purification Purified by Protein A
Form liquid
Storage Buffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Storage Temperature Store at 4°C for 12 months
Shipping Temperature 4°C
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty