Anti-GPATCH3 Rabbit Polyclonal Antibody (Cy7®)

Supplier: Bioss

BS-13500R-CY7
10299-550EA 811.1 USD
10299-550
Anti-GPATCH3 Rabbit Polyclonal Antibody (Cy7®)
Antibodies
GPATCH3

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: GPATCH3
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR