Anti-GPATCH3 Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Supplier: Bioss

BS-13500R-A350
10299-488EA 811.1 USD
10299-488
Anti-GPATCH3 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antibodies
GPATCH3

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: GPATCH3
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR