Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-FADD Rabbit Polyclonal Antibody (Biotin)

Supplier: Bioss
FADD (Fas Associated Death Domain) is an apoptosis adapter molecule enabling transduction of the apoptosis signal initiated via the FasL/Fas receptor interaction. The protein contains a C terminal death domain that interacts with the Fas receptor death domain. The N terminus contains a death effectors domain (DED) which recruits caspase to the death inducing signaling complex (DISC) and initiates the apoptotic caspase cascade. Recruitment of Caspase 8 to the Fas receptor results in oligomerization of the Caspase 8 protein, which in turn drives its autoactivation through self-cleavage. Activated Caspase 8 then activates other downstream caspases including Caspase 9, thereby commiting the cell to undergo apoptosis. FADD is implicated in non-apoptotic cellular pathways such as the regulation of cell cycle machinery in T lymphocytes. This is connected to the phosphorylation state of FADD and to the FasL/TRAIL induced transcriptional activation of cfos protooncogene. FADD also interacts with the hepatitis C virus core protein in the HEK 293 cell line.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500

Type: Primary
Antigen: FADD
Clonality: Polyclonal
Conjugation: Biotin
Host: Rabbit
Isotype: IgG
Reactivity: Rat

Log in to see your contract pricing and availability.
  Remember me on this device


Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
100 µL BS-1349R-BIO 10299-472 Each (100µl) Retrieving Restricted

Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Requires a maximum 2 day delivery.

restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page


Antigen Symbol FADD
Antigen Name Fas-Associated protein with Death Domain
Reactivity Rat
Conjugation Biotin
Clonality Polyclonal
Antibody Type Primary
Host Rabbit
Isotype IgG
Western Blot Yes
ImmunoChemistry Yes
Antigen Synonyms MORT1, FAS-associated death domain-containing protein, GIG3, MGC8528
Gene ID 266610
Modification phospho-Ser194
Concentration 1 μg/μl
Cross Adsorption No
Purification Purified by Protein A
Form liquid
Storage Buffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Storage Temperature Store at 4°C for 12 months
Shipping Temperature 4°C
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty