Anti-GPATCH2 Rabbit Polyclonal Antibody (Cy5.5®)

13499R-CY5.5
10227-190EA 559.62 USD
10227-190
Anti-GPATCH2 Rabbit Polyclonal Antibody (Cy5.5®)
GPATCH2

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: GPATCH2
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Rabbit
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR