Anti-GPAT2 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Supplier: Bioss

BS-13497R-FITC
10299-390EA 811.1 USD
10299-390
Anti-GPAT2 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antibodies
Esterifies acyl-group from acyl-ACP to the sn-1 position of glycerol-3-phosphate, an essential step in glycerolipid biosynthesis.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: GPAT2
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR