Anti-GPAA1 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Supplier: Bioss
BS-13496R-HRP
10299-376EA 810.85 USD
10299-376
Anti-GPAA1 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antibodies
Glycosylphosphatidylinositol (GPI) acts as a membrane anchor for cell surface proteins. Glycosylphosphatidylinositol anchor attachment 1 protein (GPAA1), also designated GPI anchor attachment protein 1 or GAA1 protein homolog, is a membrane protein localized to the endoplasmic reticulum which is involved in GPI-anchor biosynthesis. GPAA1 is crucial for GPI-anchoring of precursor proteins and catalyzes the attachment of GPI to proteins containing a C-terminal GPR attachment signal. GAA1 contains an N-terminal signal sequence, one cAMP- and cGMP-dependent protein kinase phosphorylation site, two potential N-glycosylation sites, one leucine zipper pattern and eight putative transmembrane domains. GPAA1 is ubiquitously expressed and shows higher levels of expression in fetal tissues than in adult tissues.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500

Type: Primary
Antigen: GPAA1
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR