Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-GOT1L1 Rabbit Polyclonal Antibody (Cy5.5®)

Supplier: Bioss
13494R-CY5.5
GOT1L1, Glutamate oxaloacetate transaminase 1-like protein 1, is a 421 amino acid member of the class-I pyridoxal-phosphate-dependent aminotransferase family. Similar to glutamate-oxaloacetate transaminase (GOT1), GOT1L1 is found primarily as a homodimer in the cytoplasmic space but also has mitochondrial and chloroplastic isozymes. GOT1L1 transaminates 2-oxoglutarate with L-aspartate to yield oxaloacetate and L-glutamate. This reaction requires a pyridoxal phosphate cofactor to occur. The GOT1L1 peptidase is predominately expressed in the liver and serum levels of this protein can be used as an indicator of liver disease. Also, elevated glutamate concentrations in the brain interstitial fluids can lead to pathological brain conditions. The glutamate-scavenging properties of these aminotranferase type enzymes likely prevent glutamate excitotoxicity and the long-lasting neurological deficits seen after stroke.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: GOT1L1
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat




Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
100 µL 13494R-CY5.5 10227-180 Each (100µl) Retrieving Restricted

Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Requires a maximum 2 day delivery.

restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen Symbol GOT1L1
Antigen Name Glutamic-oxaloacetic Transaminase 1-like 1
Reactivity Mouse, Rat, Human
Conjugation Cy5.5®
Clonality Polyclonal
Antibody Type Primary
Host Rabbit
Isotype IgG
ImmunoChemistry Yes
Antigen Synonyms Glutamic oxaloacetic transaminase 1 like 1, GOT1L1, Putative aspartate aminotransferase, Glutamate oxaloacetate transaminase 1-like protein 1, cytoplasmic 2, AATC2_HUMAN, Transaminase A-like protein 1.
Gene ID 137362
Concentration 1 μg/μl
Cross Adsorption No
Purification Purified by Protein A
Form liquid
Storage Buffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Storage Temperature Store at 4°C for 12 months
Shipping Temperature 4°C
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty