Anti-GOT1L1 Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Supplier: Bioss

BS-13494R-A647
10299-302EA 810.85 USD
10299-302
Anti-GOT1L1 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antibodies
GOT1L1, Glutamate oxaloacetate transaminase 1-like protein 1, is a 421 amino acid member of the class-I pyridoxal-phosphate-dependent aminotransferase family. Similar to glutamate-oxaloacetate transaminase (GOT1), GOT1L1 is found primarily as a homodimer in the cytoplasmic space but also has mitochondrial and chloroplastic isozymes. GOT1L1 transaminates 2-oxoglutarate with L-aspartate to yield oxaloacetate and L-glutamate. This reaction requires a pyridoxal phosphate cofactor to occur. The GOT1L1 peptidase is predominately expressed in the liver and serum levels of this protein can be used as an indicator of liver disease. Also, elevated glutamate concentrations in the brain interstitial fluids can lead to pathological brain conditions. The glutamate-scavenging properties of these aminotranferase type enzymes likely prevent glutamate excitotoxicity and the long-lasting neurological deficits seen after stroke.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: GOT1L1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR