Anti-GOLT1A Rabbit Polyclonal Antibody (Cy5.5®)

13491R-CY5.5
10227-174EA 559.62 USD
10227-174
Anti-GOLT1A Rabbit Polyclonal Antibody (Cy5.5®)
May be involved in fusion of ER-derived transport vesicles with the Golgi complex.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: GOLT1A
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR