Anti-GOLT1A Rabbit Polyclonal Antibody (Biotin)

BS-13491R-BIO
10299-238EA 529.29 USD
10299-238
Anti-GOLT1A Rabbit Polyclonal Antibody (Biotin)
May be involved in fusion of ER-derived transport vesicles with the Golgi complex.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500

Type: Primary
Antigen: GOLT1A
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR