Anti-GOLIM4 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Supplier: Bioss

BS-13490R-HRP
10299-104EA 811.1 USD
10299-104
Anti-GOLIM4 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antibodies
The Golgi complex plays a key role in the sorting and modification of proteins exported from the endoplasmic reticulum. The protein encoded by this gene is a type II Golgi-resident protein. It may process proteins synthesized in the rough endoplasmic reticulum and assist in the transport of protein cargo through the Golgi apparatus. [provided by RefSeq, Jul 2008].

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500

Type: Primary
Antigen: GOLPH4
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR